missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoaniline, 96%, pract.

Product Code. 15740869 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15740869 100 g 100g
10710482 500 g 500g
11995211 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15740869 Supplier Thermo Scientific Acros Supplier No. 402871000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-40-1 | C6H6BrN | 172.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 106-40-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.02
MDL Number MFCD00007822
InChI Key WDFQBORIUYODSI-UHFFFAOYSA-N
Synonym p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
PubChem CID 7807
IUPAC Name 4-bromoaniline
SMILES C1=CC(=CC=C1N)Br

Spezifikation

CAS Min % 95.0
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color Brown to Gray
Density 1.4970g/mL
Boiling Point 230.0°C to 250.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NH2
Quantity 100 g
Beilstein 12,636
Merck Index 15,1411
Specific Gravity 1.497
Solubility Information Solubility in water: < 1 g / l. Other solubilities: soluble in most organic solvents except cold,petroleum,very soluble in alcohol and ether
Formula Weight 172.02
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromoaniline, (pract), 96%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • 4-Bromoaniline
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: 4-Bromoaniline

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.