missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoaniline, 99+%

Product Code. 11965211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11965211 500 g Glass bottle 500g
10346450 25 g Glass bottle 25g
10764851 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11965211 Supplier Thermo Scientific Acros Supplier No. 146285000
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-40-1 | C6H6BrN | 172.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-40-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.02
InChI Key WDFQBORIUYODSI-UHFFFAOYSA-N
Synonym p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
PubChem CID 7807
IUPAC Name 4-bromoaniline
SMILES C1=CC(=CC=C1N)Br

Specifications

Melting Point 60.0°C to 64.0°C
Color Brown-Gray to White
Density 1.4970g/mL
Boiling Point 230.0°C to 250.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NH2
Quantity 500 g
Beilstein 12,636
Merck Index 15,1411
Specific Gravity 1.497
Solubility Information Solubility in water: < 1 g / l. Other solubilities: soluble in most organic solvents except cold,petroleum,very soluble in alcohol and ether
Formula Weight 172.02
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.