missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole, 98%

Product Code. 15720789 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15720789 500 mL 500mL
10254130 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15720789 Supplier Thermo Scientific Acros Supplier No. 106635000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-92-7 | C7H7BrO | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-92-7
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000097
InChI Key QJPJQTDYNZXKQF-UHFFFAOYSA-N
Synonym 4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
PubChem CID 7730
ChEBI CHEBI:47257
IUPAC Name 1-bromo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 9.0°C to 10.0°C
Color Colorless to Yellow
Density 1.4900g/mL
Boiling Point 223.0°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OCH3
Refractive Index 1.5630 to 1.5650
Quantity 500 mL
Beilstein 06, 199
Merck Index 10, 1400
Specific Gravity 1.49
Solubility Information Solubility in water: immiscible
Formula Weight 187.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromoanisole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.