missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole, 99%

Product Code. 11499413
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499413 500 g 500g
11489413 100 g 100g
11409423 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499413 Supplier Thermo Scientific Alfa Aesar Supplier No. A11824.36
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 104-92-7 | C7H7BrO | 187.036 g/mol

4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-92-7
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00000097
InChI Key QJPJQTDYNZXKQF-UHFFFAOYSA-N
Synonym 4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
PubChem CID 7730
ChEBI CHEBI:47257
IUPAC Name 1-bromo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Br

Specifications

Melting Point 10°C to 13°C
Density 1.494
Boiling Point 98°C to 99°C (10 mmHg)
Flash Point 94°C (201°F)
Assay Percent Range 99%
Refractive Index 1.564
Quantity 500 g
Beilstein 1237590
Merck Index 14,1427
Solubility Information Immiscible with water.
Formula Weight 187.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromoanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.