missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals

Product Code. 11450608
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450608 100 g 100g
11446257 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450608 Supplier Thermo Scientific Alfa Aesar Supplier No. B25163.22
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34421-94-8 | Molecular Formula: C11H15BrO2 | Molecular Weight: 259.14 g/mol

4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 34421-94-8
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01863514
InChI Key BFSNEBVTOODGHZ-UHFFFAOYSA-N
Synonym 4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
PubChem CID 688340
IUPAC Name 1-bromo-4-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=C(Br)C=C1

Spécification

Density 1.274
Boiling Point 99°C to 101°C (1 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.512
Quantity 100 g
Beilstein 2448775
Formula Weight 259.14
Percent Purity 98%
Chemical Name or Material 4-Bromobenzaldehyde diethyl acetal

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.