missing translation for 'onlineSavingsMsg'
Learn More

4-bromobenzaldehyde, Thermo Scientific™

Product Code. 10075602
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10075602 10 g 10g
10080364 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10075602 Supplier Thermo Scientific Maybridge Supplier No. SB01132EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1122-91-4
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
InChI Key ZRYZBQLXDKPBDU-UHFFFAOYSA-N
Synonym p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
PubChem CID 70741
IUPAC Name 4-bromobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Br

Specifications

Assay Percent Range 95%
Linear Formula BrC6H4CHO
Quantity 10 g
Formula Weight 185.02
Percent Purity 95%
Chemical Name or Material 4-Bromobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.