missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzhydrazide, 98+%

Product Code. 11423168
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11423168

Brand: Thermo Scientific Alfa Aesar A18918.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5933-32-4 | C7H7BrN2O | 215.05 g/mol

4-Bromobenzoic hydrazide has been used in the preparation of {2-[1-oxo-3-(4-bromophenyl)-2-propenyl]hydrazide}-4-bromobenzoic acid, {2,2?-[1,4-phenylenebis(1-oxo-2-propene-3,1-diyl)]dihydrazide}-p-bromobenzoic acid and substituted period[1,2-a]pyrimidine-2,4(3H)-diones (PPMDO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzoic hydrazide has been used in the preparation of {2-[1-oxo-3-(4-bromophenyl)-2-propenyl]hydrazide}-4-bromobenzoic acid, {2,2′-[1,4-phenylenebis(1-oxo-2-propene-3,1-diyl)]dihydrazide}-p-bromobenzoic acid and substituted period[1,2-a]pyrimidine-2,4(3H)-diones (PPMDO).

Solubility
Soluble in hot methanol (almost transparency). Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5933-32-4
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00007602
InChI Key UYIMBYKIIMYFPS-UHFFFAOYSA-N
Synonym 4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine
PubChem CID 22219
IUPAC Name 4-bromobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)Br

Specifications

Melting Point 167°C to 169°C
Quantity 5 g
Beilstein 777006
Solubility Information Soluble in hot methanol (almost transparency). Slightly soluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzhydrazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.