missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoic acid, 97%

Product Code. 10654812 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10654812 25 g Plastic bottle 25g
10376740 100 g Plastic bottle 100g
10157643 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10654812 Supplier Thermo Scientific Acros Supplier No. 148700250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 586-76-5 | C7H5BrO2 | 201.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-76-5
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00002529
InChI Key TUXYZHVUPGXXQG-UHFFFAOYSA-N
Synonym p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acid
PubChem CID 11464
ChEBI CHEBI:60698
IUPAC Name 4-bromobenzoic acid
SMILES OC(=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 251°C to 256°C
Color Brown-Pink to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula BrC6H4CO2H
Quantity 25 g
Beilstein 09, 351
Merck Index 15, 1415
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 201.02
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 4-Bromobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.