missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoic Acid Methyl Ester, TRC

Product Code. 17953145
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953145 100 g 100g
17914395 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17953145 Supplier Toronto Research Chemicals Supplier No. TRCB680245100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-42-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
Synonym 1-Bromo-4-(methoxycarbonyl)benzene, 4-Methoxycarbonyl-1-bromobenzene, 4-Methoxycarbonylphenyl Bromide, Methyl 4-Bromobenzoate, Methyl p-Bromobenzoate, NSC 9392, p-Bromobenzoic Acid Methyl Ester;
IUPAC Name methyl 4-bromobenzoate
SMILES COC(=O)c1ccc(Br)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H7BrO2/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5H,1H3
Formula Weight 213.9629
Chemical Name or Material 4-Bromobenzoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.