missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzophenone, 98%

Product Code. 11389409
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389409 25 g 25g
11399409 100 g 100g
11457806 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389409 Supplier Thermo Scientific Alfa Aesar Supplier No. A14755.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-90-4 | C13H9BrO | 261.12 g/mol

4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

Solubility
Insoluble in water but slightly soluble in other solvents such as ethanol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-90-4
Molecular Formula C13H9BrO
Molecular Weight (g/mol) 261.12
MDL Number MFCD00000103
InChI Key KEOLYBMGRQYQTN-UHFFFAOYSA-N
Synonym 4-bromobenzophenone, 4-bromophenyl phenyl methanone, p-bromobenzophenone, methanone, 4-bromophenyl phenyl, benzophenone, 4-bromo, p-benzoylbromobenzene, usaf do-3, 4-bromophenyl phenyl ketone, 4-bromophenyl-phenyl-methanone, 4-bromophenyl-phenylmethanone
PubChem CID 7030
SMILES BrC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 79°C to 83°C
Boiling Point 350°C
Odor Odorless
Quantity 25 g
Beilstein 1910182
Solubility Information Insoluble in water but slightly soluble in other solvents such as ethanol,ether and benzene.
IUPAC Name (4-bromophenyl)-phenylmethanone
Formula Weight 261.12
Percent Purity 98%
Chemical Name or Material 4-Bromobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.