missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylamine hydrochloride, 98%, ACROS Organics™

Product Code. 10606782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10606782 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10606782 Supplier Thermo Scientific Acros Supplier No. 194650050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 26177-44-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 272°C to 276°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H9BrClN
Linear Formula BrC6H4CH2NH2·HCl
MDL Number MFCD00012860
Quantity 5 g
Beilstein 12, 1075
Synonym 4-bromobenzylamine hydrochloride, 4-bromophenyl methanamine hydrochloride, 4-bromobenzylamine hcl, 4-bba x hcl, 4-bromo-benzylamine hydrochloride, p-bromobenzylamine hydrochloride, benzenemethanamine, 4-bromo-, hydrochloride, pubchem3751, acmc-1cfrz, 4-bromobenzylamine, hcl
InChI Key BVYUYDBWQMSOKM-UHFFFAOYSA-N
SMILES [H+].[Cl-].NCC1=CC=C(Br)C=C1
IUPAC Name hydrogen 1-(4-bromophenyl)methanamine chloride
Molecular Weight (g/mol) 222.51
PubChem CID 2724096
Formula Weight 222.52
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzylamine hydrochloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.