Learn More
4-Bromobiphenyl, 98+%
Description
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.
Solubility
Insoluble in water.
Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
Specifications
Spécification
| Melting Point | 88°C to 92°C |
| Boiling Point | 310°C |
| Flash Point | 143°C (289°F) |
| Quantity | 10 g |
| Beilstein | 1907453 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 233.12 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Bromobiphenyl |
Sécurité et traitement
- 4-Bromobiphenyl
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: 4-Bromodiphenyl
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.