missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobiphenyl, 98+%

Product Code. p-73164 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10482581

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-66-0 | C12H9Br | 233.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-66-0
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000100
InChI Key PKJBWOWQJHHAHG-UHFFFAOYSA-N
Synonym 4-bromobiphenyl, p-bromobiphenyl, 4-bromo-1,1'-biphenyl, 4-bromodiphenyl, 1,1'-biphenyl, 4-bromo, 4-biphenyl bromide, p-bromodiphenyl, biphenyl, 4-bromo, 4-bromo-biphenyl, p-phenylbromobenzene
PubChem CID 7101
IUPAC Name 1-bromo-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 89.0°C to 92.0°C
Color White to Yellow
Boiling Point 310.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5C6H4Br
Quantity 25 g
Beilstein 05, 580
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, ether and benzene
Formula Weight 233.11
Percent Purity 99%
Physical Form Crystalline Flakes
Chemical Name or Material 4-Bromobiphenyl, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Bromobiphenyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.