missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromobutyl)triphenylphosphonium bromide, 98%

Product Code. 11486226
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11486226

Brand: Thermo Scientific Alfa Aesar B23040.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7333-63-3 | C22H23Br2P | 478.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7333-63-3
Molecular Formula C22H23Br2P
Molecular Weight (g/mol) 478.21
MDL Number MFCD00031636
InChI Key BJDNCJGRAMGIRU-UHFFFAOYSA-M
Synonym 4-bromobutyl triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphonium bromide, 4-bromobutyl triphenylphosphanium bromide, 4-bromobutyl-triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphanium bromide, 4-bromobutyl triphenyl-phosphonium bromide, 4-bromo butyl triphenylphosphonium bromide, 4-bromanylbutyl triphenyl phosphanium bromide, phosphonium, 4-bromobutyl triphenyl-,bromide 1:1, phosphonium, 4-bromobutyl triphenyl-, bromide 1:1
PubChem CID 2733851
IUPAC Name 4-bromobutyl(triphenyl)phosphanium;bromide
SMILES [Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 206°C to 212°C
Quantity 50 g
Beilstein 3812291
Sensitivity Hygroscopic
Formula Weight 478.22
Percent Purity 98%
Chemical Name or Material (4-Bromobutyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.