missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocinnamic acid, predominantly trans, 97+%

Product Code. 11440423
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440423 100 g 100g
11420423 5 g 5g
11430423 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440423 Supplier Thermo Scientific Alfa Aesar Supplier No. A11503.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1200-07-3 | C9H7BrO2 | 227.06 g/mol

4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

Solubility
Soluble in alcohol, ethyl acetate, DMSO. Insoluble in water.

Notes
Store at -20°C. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1200-07-3
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00004394
InChI Key CPDDDTNAMBSPRN-ZZXKWVIFSA-N
Synonym 4-bromocinnamic acid, p-bromocinnamic acid, 3-4-bromophenyl acrylic acid, bromocinnamic acid,4, 2e-3-4-bromophenyl prop-2-enoic acid, trans-4-bromocinnamic acid, e-3-4-bromophenyl acrylic acid, 2-propenoic acid, 3-4-bromophenyl-, 2e, 4-bromocinnamic acid, predominantly trans, rarechem bk hd c006
PubChem CID 737158
IUPAC Name (E)-3-(4-bromophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=C(Br)C=C1

Specifications

Melting Point 260°C to 264°C
Odor Odorless
Quantity 100 g
Beilstein 3538823
Solubility Information Soluble in alcohol,ethyl acetate,DMSO. Insoluble in water.
Formula Weight 227.06
Percent Purity ≥98%
Chemical Name or Material 4-Bromocinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.