missing translation for 'onlineSavingsMsg'
Learn More

4-Bromofluorobenzene, 99%

Product Code. 10385830 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10385830 10 mL Glass bottle 10mL
10345880 100 mL Glass bottle 100mL
15730789 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10385830 Supplier Thermo Scientific Supplier No. 107010100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 460-00-4 | C6H4BrF | 175.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 460-00-4
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175.00
MDL Number MFCD00000342
InChI Key AITNMTXHTIIIBB-UHFFFAOYSA-N
Synonym 4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
PubChem CID 9993
IUPAC Name 1-bromo-4-fluorobenzene
SMILES FC1=CC=C(Br)C=C1

Specifikationer

CAS Min % 98.5
CAS Max % 100.0
Melting Point -8.0°C
Color Colorless
Density 1.5930g/mL
Boiling Point 150.0°C to 153.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4F
Refractive Index 1.5260 to 1.5280
Quantity 10 mL
Beilstein 05, 209
Specific Gravity 1.593
Solubility Information Solubility in water: 0.14g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,acetone,soluble in ether
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromofluorobenzene, 99%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.