missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomandelic acid, 96%, Thermo Scientific™

Product Code. 10626482 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 10626482

Brand: Thermo Scientific Acros 107110100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6940-50-7
CAS Min % 95.0
CAS Max % 100.0
Melting Point 115.0°C to 120.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H7BrO3
Linear Formula BrC6H4CH(OH)COOH
MDL Number MFCD00004232
Quantity 10 g
Beilstein 10, 210
Merck Index 15, 1434
Synonym 4-bromomandelic acid, 2-4-bromophenyl-2-hydroxyacetic acid, p-bromomandelic acid, p-bromo mandelic acid, 4-bromo-dl-mandelic acid, benzeneacetic acid, 4-bromo-.alpha.-hydroxy, 4-bromophenyl hydroxy acetic acid, 4-bromo-alpha-hydroxyphenylacetic acid, 4-bromo-hydroxybenzeneaceticacid, mandelic acid, p-bromo
Solubility Information Solubility in water: slightly soluble
InChI Key BHZBRPQOYFDTAB-UHFFFAOYNA-N
SMILES OC(C(O)=O)C1=CC=C(Br)C=C1
IUPAC Name 2-(4-bromophenyl)-2-hydroxyacetic acid
Molecular Weight (g/mol) 231.05
PubChem CID 97930
Formula Weight 231.05
Percent Purity 96%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-Bromomandelic acid, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromomandelic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.