missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-3-nitrobenzoic acid, 97%

Product Code. 11460798
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11460798

Brand: Thermo Scientific Alfa Aesar B25255.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55715-03-2 | C8H6BrNO4 | 260.043 g/mol

4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.

Solubility
Soluble in DMF and dichloromethane. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55715-03-2
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.043
MDL Number MFCD00216575
InChI Key QMAHVAFURJBOFV-UHFFFAOYSA-N
Synonym 4-bromomethyl-3-nitrobenzoic acid, 4-bromomethyl-3-nitrobenzoicacid, benzoic acid, 4-bromomethyl-3-nitro, 3-nitro-4-bromomethylbenzoic acid, bnba, acmc-1auor, ksc495i9f, 3-nitro-4-bromomethyl benzoic acid, 4-bromomethyl-3-nitro benzoic acid
PubChem CID 132386
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])CBr

Specifications

Melting Point 128°C to 133°C
Quantity 1 g
UN Number UN3261
Solubility Information Soluble in DMF and dichloromethane. Insoluble in water.
Formula Weight 260.05
Percent Purity 97%
Chemical Name or Material 4-Bromomethyl-3-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.