missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-6,7-dimethoxycoumarin, 95%

Product Code. 15471897
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
Unit Size:
0.1g
0.5g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471897 0.1 g 0.1g
15451897 0.5 g 0.5g
15461897 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471897 Supplier Thermo Scientific Alfa Aesar Supplier No. 043402.77
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorescent labeling reagent; Used in HPLC for the detection of carboxylic acids | CAS: 88404-25-5 | C12H11BrO4 | 299.12 g/mol

4-Bromomethyl-6,7-dimethoxycoumarin is used as fluorescent labeling reagent for the reversed-phase HPLC separation and detection of carboxylic acids. It is also used as derivatization reagent: for quantitation of clofibric, bezafibric and fenofibric acids in liver by liquid chromatography, in determination of the concentration of tripeptide glutathione by matrix-assisted laser desorption ionization time-of-flight mass spectrometry and for determination of carboxylic acids in chromatographic detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88404-25-5
Molecular Formula C12H11BrO4
Molecular Weight (g/mol) 299.12
MDL Number MFCD00011570
InChI Key JGODLBJJCNQFII-UHFFFAOYSA-N
Synonym 4-bromomethyl-6,7-dimethoxycoumarin, 4-bromomethyl-6,7-dimethoxy-2h-chromen-2-one, 4-bromomethyl-6,7-dimethoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-6,7-dimethoxy, 4-bromomethyl-6,7-dimethoxy coumarin, 4-bdmc, brdmc-4,6,7, 4-bromomethyl-6,7-dimethoxy-2-oxo-2h-benzopyran
PubChem CID 128870
IUPAC Name 4-(bromomethyl)-6,7-dimethoxychromen-2-one
SMILES COC1=C(C=C2C(=C1)C(=CC(=O)O2)CBr)OC

Specifications

Melting Point 200°C to 203°C
Quantity 0.1 g
UN Number UN3261
Beilstein 4750487
Solubility Information Insoluble in water.
Formula Weight 299.12
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Bromomethyl-6,7-dimethoxycoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.