missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10682875
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10682875 250 mg 250mg
10581724 1 g 1g
10517484 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10682875 Supplier Thermo Scientific Maybridge Supplier No. MO07312CB
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51359-78-5
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
InChI Key XYPVBKDHERGKJG-UHFFFAOYSA-N
PubChem CID 11206421
IUPAC Name 4-(bromomethyl)benzaldehyde
SMILES C1=CC(=CC=C1CBr)C=O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Synonym 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde
Formula Weight 199.05
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.