missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzoic acid, 97%

Product Code. 11302126
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302126 25 g 25g
11392116 5 g 5g
11312126 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302126 Supplier Thermo Scientific Alfa Aesar Supplier No. L03371.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6232-88-8 | C8H7BrO2 | 215.046 g/mol

4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.

Solubility
Soluble in water.

Notes
Incompatible with bases, amines and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6232-88-8
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00002567
InChI Key CQQSQBRPAJSTFB-UHFFFAOYSA-N
Synonym 4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid
PubChem CID 22599
IUPAC Name 4-(bromomethyl)benzoic acid
SMILES C1=CC(=CC=C1CBr)C(=O)O

Specifications

Melting Point 223°C to 227°C
Quantity 25 g
UN Number UN3261
Beilstein 1862870
Solubility Information Soluble in water.
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.