missing translation for 'onlineSavingsMsg'
Learn More

4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%

Product Code. 15411618
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411618 5 g 5g
15401618 1 g 1g
15421618 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411618 Supplier Thermo Scientific Alfa Aesar Supplier No. H55394.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 114772-54-2 | C14H10BrN | 272.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-54-2
Molecular Formula C14H10BrN
Molecular Weight (g/mol) 272.145
MDL Number MFCD00671503
InChI Key LFFIEVAMVPCZNA-UHFFFAOYSA-N
Synonym 4'-bromomethyl-2-cyanobiphenyl, 2-4-bromomethyl phenyl benzonitrile, 2-cyano-4'-bromomethylbiphenyl, 4'-bromomethyl-1,1'-biphenyl-2-carbonitrile, 2-4-bromo-methylphenyl benzonitrile, 4'-bromomethyl-2-biphenylcarbonitrile, 4'-bromomethylbiphenyl-2-carbonitrile, 2'-cyano-4-bromomethylbiphenyl, 4-2-cyanophenyl benzyl bromide, unii-ui5jr3k10f
PubChem CID 1501912
IUPAC Name 2-[4-(bromomethyl)phenyl]benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
UN Number UN3439
Formula Weight 272.14
Percent Purity 98%
Chemical Name or Material 4'-(Bromomethyl)biphenyl-2-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.