missing translation for 'onlineSavingsMsg'
Learn More

4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%

Product Code. p-7036922
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15401618

Brand: Thermo Scientific Alfa Aesar H55394.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 114772-54-2 | C14H10BrN | 272.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-54-2
Molecular Formula C14H10BrN
Molecular Weight (g/mol) 272.145
MDL Number MFCD00671503
InChI Key LFFIEVAMVPCZNA-UHFFFAOYSA-N
Synonym 4'-bromomethyl-2-cyanobiphenyl, 2-4-bromomethyl phenyl benzonitrile, 2-cyano-4'-bromomethylbiphenyl, 4'-bromomethyl-1,1'-biphenyl-2-carbonitrile, 2-4-bromo-methylphenyl benzonitrile, 4'-bromomethyl-2-biphenylcarbonitrile, 4'-bromomethylbiphenyl-2-carbonitrile, 2'-cyano-4-bromomethylbiphenyl, 4-2-cyanophenyl benzyl bromide, unii-ui5jr3k10f
PubChem CID 1501912
IUPAC Name 2-[4-(bromomethyl)phenyl]benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr

Specifications

Melting Point 125°C to 128°C
Quantity 1 g
UN Number UN3439
Formula Weight 272.14
Percent Purity 98%
Chemical Name or Material 4'-(Bromomethyl)biphenyl-2-carbonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.