missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenoxyacetic acid, 95%, Thermo Scientific™

Product Code. 15417398
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417398 25 g 25g
15407398 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417398 Supplier Thermo Scientific Alfa Aesar Supplier No. H60888.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-91-7
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00093070
InChI Key SZEBGAQWWSUOHT-UHFFFAOYSA-N
Synonym 4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy
PubChem CID 74657
IUPAC Name 2-(4-bromophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)Br

Specifications

Melting Point 149°C to 153°C
Quantity 25 g
Formula Weight 231.04
Percent Purity 95%
Chemical Name or Material 4-Bromophenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.