missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl methyl sulfone, 98+%

Product Code. 11430273
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430273 10 g 10g
11440273 50 g 50g
11450273 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430273 Supplier Thermo Scientific Alfa Aesar Supplier No. A11427.09
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3466-32-8 | C7H7BrO2S | 235.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3466-32-8
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.095
MDL Number MFCD00025065
InChI Key FJLFSYRGFJDJMQ-UHFFFAOYSA-N
Synonym 4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl
PubChem CID 77014
IUPAC Name 1-bromo-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 101°C to 102°C
Quantity 10 g
Beilstein 2045675
Formula Weight 235.1
Percent Purity ≥98%
Chemical Name or Material 4-Bromophenyl methyl sulfone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.