missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenyl)urea, TRC

Product Code. 17943895
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943895 500 mg 500mg
17992395 100 mg 100mg
17990605 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943895 Supplier Toronto Research Chemicals Supplier No. TRCB700233500MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1967-25-5
Molecular Formula C7 H7 N2 O Br
Molecular Weight (g/mol) 215.05
Synonym N-(4-Bromophenyl)urea (ACI), Urea, (4-bromophenyl)- (9CI), Urea, (p-bromophenyl)- (6CI, 7CI, 8CI), (p-Bromophenyl)urea, 1-(4-Bromophenyl)urea, 1-(p-Bromophenyl)urea, 4-Bromophenylurea, Urea, N-(4-bromophenyl)- (ACI)
IUPAC Name (4-bromophenyl)urea
SMILES NC(=O)Nc1ccc(Br)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H7BrN2O/c8-5-1-3-6(4-2-5)10-7(9)11/h1-4H,(H3,9,10,11)
Formula Weight 213.9742
Percent Purity >95%
Chemical Name or Material (4-Bromophenyl)urea
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.