missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetone, 97%

Product Code. 10404273 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10404273

Brand: Thermo Scientific Acros 380220050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6186-22-7 | C24H24N2O5S | 452.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6186-22-7
Molecular Formula C24H24N2O5S
Molecular Weight (g/mol) 452.53
MDL Number MFCD00210401
InChI Key CDUPPXGKUKTGLU-UHFFFAOYSA-N
Synonym 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone
PubChem CID 736332
SMILES COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5560 to 1.5590 (20°C, 589nm)
Quantity 5 g
IUPAC Name 1-(4-bromophenyl)propan-2-one
Formula Weight 213.08
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 4-Bromophenylacetone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.