missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetone, 98%

Product Code. 11499326
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11499326

Brand: Thermo Scientific Alfa Aesar A19028.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6186-22-7 | C24H24N2O5S | 452.53 g/mol

4-Bromophenylacetone is used to prepare biphenyl-4-yl-acetone by reacting with phenylboronic acid using cesium fluoride as a reagent and palladium phosphine complex as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenylacetone is used to prepare biphenyl-4-yl-acetone by reacting with phenylboronic acid using cesium fluoride as a reagent and palladium phosphine complex as a catalyst.

Solubility
Miscible with chloroform and dichloromethane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6186-22-7
Molecular Formula C24H24N2O5S
Molecular Weight (g/mol) 452.53
MDL Number MFCD00210401
InChI Key CDUPPXGKUKTGLU-UHFFFAOYSA-N
Synonym 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone
PubChem CID 736332
IUPAC Name 1-(4-bromophenyl)propan-2-one
SMILES COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1

Specifications

Quantity 25 g
Solubility Information Miscible with chloroform and dichloromethane. Slightly miscible with water.
Formula Weight 213.07
Percent Purity 98%
Chemical Name or Material 4-Bromophenylacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.