missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylhydrazine hydrochloride, 99%

Product Code. 10571411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571411 10 g Glass bottle 10g
10581411 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10571411 Supplier Thermo Scientific Acros Supplier No. 162310100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 622-88-8 | C6H7BrN2·HCl | 223.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-88-8
Molecular Formula C6H7BrN2·HCl
Molecular Weight (g/mol) 223.51
MDL Number MFCD00012941
InChI Key RGGOWBBBHWTTRE-UHFFFAOYSA-N
Synonym 4-bromophenylhydrazine hydrochloride, 4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazine hcl, 4-bromo phenyl hydrazine hydrochloride, p-bromophenylhydrazine monohydrochloride, hydrazine, 4-bromophenyl-, monohydrochloride, 1-4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazinehydrochloride, p-bromophenylhydrazine hydrochloride
PubChem CID 12157
IUPAC Name (4-bromophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Br.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 210°C to 215°C
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4NHNH2·HCl
Quantity 10 g
Beilstein 15, 434
Solubility Information Solubility in water: soluble
Formula Weight 223.51
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenylhydrazine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromophenylhydrazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.