missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromostyrene oxide, 96%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 32017-76-8 |
| CAS Min % | 95.0 |
| CAS Max % | 100.0 |
| Melting Point | 26.0°C to 27.0°C |
| Color | White to Yellow |
| Density | 1.5040g/mL |
| Boiling Point | 91.0°C to 93.0°C (2.0 mmHg) |
| Flash Point | 102°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (GC) |
| Show More |
Safety and Handling
Product Identifier
- 4-Bromostyrene oxide
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction