missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothioanisole, 97%

Product Code. 10477570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477570 25 g Glass bottle 25g
10611453 5 g Glass bottle 5g
10357750 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10477570 Supplier Thermo Scientific Acros Supplier No. 188190250
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 104-95-0 | C7H7BrS | 203.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-95-0
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD00000102
InChI Key YEUYZNNBXLMFCW-UHFFFAOYSA-N
Synonym 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole
PubChem CID 66037
IUPAC Name 1-bromo-4-methylsulfanylbenzene
SMILES CSC1=CC=C(Br)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 35.0°C to 40.0°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4SCH3
Quantity 25 g
Beilstein 06, 330
Formula Weight 203.1
Percent Purity 97%
Physical Form Low Melting Crystalline Mass, Crystals, Crystalline Powder or Chunks
Chemical Name or Material 4-Bromothioanisole, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.