missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothioanisole, 98+%

Product Code. 11472897
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472897 100 g 100g
11482897 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472897 Supplier Thermo Scientific Alfa Aesar Supplier No. A15504.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 104-95-0 | C7H7BrS | 203.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-95-0
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD00000102
InChI Key YEUYZNNBXLMFCW-UHFFFAOYSA-N
Synonym 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole
PubChem CID 66037
IUPAC Name 1-bromo-4-methylsulfanylbenzene
SMILES CSC1=CC=C(Br)C=C1

Specifications

Melting Point 37°C to 40°C
Boiling Point 250°C
Flash Point >110°C (230°F)
Odor Stench
Quantity 100 g
Beilstein 1906693
Formula Weight 203.11
Percent Purity ≥98%
Chemical Name or Material 4-Bromothioanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.