missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophenol, 95%

Product Code. 10214710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10214710 100 g Glass bottle 100g
10028591 5 g Glass bottle 5g
10020730 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10214710 Supplier Thermo Scientific Acros Supplier No. 107461000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-53-6
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004845
InChI Key FTBCOQFMQSTCQQ-UHFFFAOYSA-N
Synonym 4-bromothiophenol, benzenethiol, 4-bromo, p-bromothiophenol, p-bromobenzenethiol, 4-bromobenzene-1-thiol, 4-bromo-benzenethiol, 4-bromo thiophenol, p-bromophenyl thiol, benzenethiol, p-bromo, unii-p3wuo03x17
PubChem CID 66049
IUPAC Name 4-bromobenzenethiol
SMILES C1=CC(=CC=C1S)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 71°C to 76°C
Color White to Beige
Boiling Point 239°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula BrC6H4SH
Quantity 100 g
Beilstein 06, 330
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alkaline solutions (reacts),soluble in methanol
Formula Weight 189.07
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromothiophenol, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.