missing translation for 'onlineSavingsMsg'
Learn More

4-Bromotoluene, 99%

Product Code. 10326170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10326170 500 mL Glass bottle 500mL
10243940 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10326170 Supplier Thermo Scientific Supplier No. 107485000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-38-7 | C7H7Br | 171.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-38-7
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000109
InChI Key ZBTMRBYMKUEVEU-UHFFFAOYSA-N
Synonym 4-bromotoluene, p-bromotoluene, 4-methylbromobenzene, p-tolyl bromide, benzene, 1-bromo-4-methyl, toluene, p-bromo, p-methylphenyl bromide, p-methylbromobenzene, 1-methyl-4-bromobenzene, 4-bromo-1-methylbenzene
PubChem CID 7805
IUPAC Name 1-bromo-4-methylbenzene
SMILES CC1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 26°C to 29°C
Color White
Density 1.3900g/mL
Boiling Point 184°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4Br
Refractive Index 1.549
Quantity 500 mL
Beilstein 05, 305
Merck Index 14, 1439
Specific Gravity 1.39
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 171.04
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Bromotoluene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromotoluene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.