missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-1,2-difluorobenzene, 97%, Thermo Scientific™

Product Code. 10748153
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10748153 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10748153 Supplier Thermo Scientific Maybridge Supplier No. SB01824EA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 696-02-6
CAS Min % 97.0
Density 1.35g/mL
Assay Percent Range 97%
Molecular Formula C6H3ClF2
MDL Number MFCD00042572
Quantity 10 g
Synonym 1-chloro-3,4-difluorobenzene, 3,4-difluorochlorobenzene, 4-chloro-1,2-difluoro-benzene, benzene, 4-chloro-1,2-difluoro, pubchem3417, 3,4-diflurochlorobenzene, acmc-209o9n, ksc493g2d, 3,4-difluoro-1-chlorobenzene
InChI Key OPQMRQYYRSTBME-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)F)F
IUPAC Name 4-chloro-1,2-difluorobenzene
Molecular Weight (g/mol) 148.537
PubChem CID 136519
Formula Weight 148.54
Percent Purity 97%
Chemical Name or Material 4-Chloro-1, 2-difluorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.