missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-naphthol, 97%

Product Code. 10560171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10560171 5 g Glass bottle 5g
10264130 25 g Glass bottle 25g
15750789 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10560171 Supplier Thermo Scientific Acros Supplier No. 109550050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 604-44-4 | C10H7ClO | 178.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-44-4
Molecular Formula C10H7ClO
Molecular Weight (g/mol) 178.62
MDL Number MFCD00003974
InChI Key LVSPDZAGCBEQAV-UHFFFAOYSA-N
Synonym 4-chloro-1-naphthol, 1-chloro-4-hydroxynaphthalene, 1-naphthalenol, 4-chloro, 4-chloro-1-naphthalenol, 1-naphthol, 4-chloro, 4-chloronaphthol, 4-chloro-.alpha.-naphthol, 4-chloro-1-hydroxynaphthalene, 4-chloro-alpha-naphthol, acmc-20amm7
PubChem CID 11787
IUPAC Name 4-chloronaphthalen-1-ol
SMILES C1=CC=C2C(=C1)C(=CC=C2Cl)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 117°C to 120°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC10H6OH
Quantity 5 g
Beilstein 06, 611
Solubility Information Solubility in water: negligible in water. Other solubilities: soluble in methanol and 8% 2,2′oxydiethanol
Formula Weight 178.62
Percent Purity 97%
Physical Form Needles or Powder
Chemical Name or Material 4-Chloro-1-naphthol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-1-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Chloro-1-naphthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.