missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-dimethoxyaniline, 98%

Product Code. 11376836
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376836 100 g 100g
11366836 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376836 Supplier Thermo Scientific Alfa Aesar Supplier No. L04202.22
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6358-64-1 | C8H10ClNO2 | 187.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6358-64-1
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.62
MDL Number MFCD00014893
InChI Key YGUFQYGSBVXPMC-UHFFFAOYSA-N
Synonym 2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718
PubChem CID 22833
IUPAC Name 4-chloro-2,5-dimethoxyaniline
SMILES COC1=CC(Cl)=C(OC)C=C1N

Specifications

Melting Point 118°C to 121°C
Flash Point 163°C (325°F)
Quantity 100 g
Beilstein 880445
Formula Weight 187.63
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,5-dimethoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.