missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-dimethylphenol, 98%

Product Code. 11440234
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440234 10 g 10g
11450234 50 g 50g
11460234 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440234 Supplier Thermo Scientific Alfa Aesar Supplier No. A13649.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1123-63-3 | C8H9ClO | 156.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-63-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00020138
InChI Key VWYKSJIPZHRLNO-UHFFFAOYSA-N
PubChem CID 70747
IUPAC Name 4-chloro-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)Cl

Specifications

Melting Point 80°C to 83°C
Quantity 10 g
Beilstein 1908282
Synonym 4-chloro-2,6-xylenol, phenol, 4-chloro-2,6-dimethyl, 2,6-dimethyl-4-chlorophenol, 4-chloro-2,6-dimethyl-phenol, 2,6-xylenol, 4-chloro, pubchem2657, acmc-1c4bc, phenol,4-chloro-2,6-dimethyl, 4-chloro-2,6-xylenol oh=1, 4-chloro-2,6-dimethylphenol
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,6-dimethylphenol
compliance-icons
Product Identifier
  • 4-Chloro-2,6-dimethylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Chloro-2,6-xylenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.