missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylaniline, 98%

Product Code. p-2690613 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10212702

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-69-2 | C7H8ClN | 141.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-69-2
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.6
MDL Number MFCD00007842
InChI Key CXNVOWPRHWWCQR-UHFFFAOYSA-N
Synonym 4-chloro-o-toluidine, 2-amino-5-chlorotoluene, fast red tr base, p-chloro-o-toluidine, kambamine red tr, benzenamine, 4-chloro-2-methyl, red tr base, 5-chloro-2-aminotoluene, 2-methyl-4-chloroaniline, 4-chloro-2-toluidine
PubChem CID 7251
ChEBI CHEBI:82276
IUPAC Name 4-chloro-2-methylaniline
SMILES CC1=C(C=CC(=C1)Cl)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 30.0°C
Density 1.1900g/mL
Boiling Point 241.0°C
Flash Point 124°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(CH3)NH2
Refractive Index 1.5845 to 1.5865
Quantity 5 g
Beilstein 12,835
Specific Gravity 1.19
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methylaniline, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-2-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.