missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-2-methylbenzoic acid, 97%, Thermo Scientific™

Product Code. p-2701668 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10395492

Brand: Thermo Scientific Maybridge TL00910DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7499-07-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045852
InChI Key XXFKOBGFMUIWDH-UHFFFAOYSA-N
Synonym 4-chloro-2-methylbenzoicacid, 4-chloro-o-toluic acid, benzoic acid, 4-chloro-2-methyl, 2-methyl-4-chlorobenzoic acid, 4-chloro-2-methyl-benzoic acid, pubchem4570, acmc-1bjnb, o-toluic acid, 4-chloro, ksc497q4r, 4-chloro-2-methylbenzoic acid
PubChem CID 348269
IUPAC Name 4-chloro-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Cl)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylbenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.