missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrotoluene, 99%

Product Code. 11410997
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410997 500 g 500g
11400997 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410997 Supplier Thermo Scientific Alfa Aesar Supplier No. B23382.36
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-59-8 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-59-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007215
InChI Key SQFLFRQWPBEDHM-UHFFFAOYSA-N
Synonym 4-chloro-2-nitrotoluene, 2-nitro-4-chlorotoluene, benzene, 4-chloro-1-methyl-2-nitro, 4,2-chloronitrotoluene, p-chloro-o-nitrotoluene, toluene, 4-chloro-2-nitro, 4-chloro-1-methyl-2-nitro-benzene, ccris 3116, 4-chlor-2-nitrotoluol, pubchem18919
PubChem CID 6975
IUPAC Name 4-chloro-1-methyl-2-nitrobenzene
SMILES CC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 34°C to 38°C
Boiling Point 242°C
Flash Point 120°C (248°F)
Quantity 500 g
UN Number UN3457
Formula Weight 171.58
Percent Purity 99%
Chemical Name or Material 4-Chloro-2-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.