missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11362489
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11362489

Brand: Thermo Scientific Alfa Aesar H26620.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-2-(trifluoromethyl)benzyl bromide is used in the synthesis of novel series of dithiocarbamates, via reaction with sodium salts of N,N-disubstituted dithiocarbamic acids

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886496-75-9
Molecular Formula C8H5BrClF3
Molecular Weight (g/mol) 273.477
MDL Number MFCD04115840
InChI Key LFFCKOROEHFKQA-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-chloro-2-trifluoromethyl benzene, 4-chloro-2-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-4-chloro-2-trifluoromethyl, 1-bromomethyl-4-chloro-2-trifluoromethyl-benzene, 4-chloro-2-trifluoromethylbenzyl bromide
PubChem CID 3492597
IUPAC Name 1-(bromomethyl)-4-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)CBr

Specifications

Refractive Index 1.52
Quantity 5 g
UN Number UN3265
Solubility Information Difficult to mix.
Formula Weight 273.48
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.