missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%

Product Code. 11352297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352297 1 g 1g
11362297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352297 Supplier Thermo Scientific Alfa Aesar Supplier No. L09850.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16588-69-5 | C8H3ClF3NO | 221.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16588-69-5
Molecular Formula C8H3ClF3NO
Molecular Weight (g/mol) 221.56
MDL Number MFCD00013873
InChI Key SBDPJLJFODHSFF-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethyl phenyl isocyanate, 4-chloro-1-isocyanato-2-trifluoromethyl benzene, 4-chloro-2-trifluoromethyl phenylisocyanate, 5-chloro-2-isocyanatobenzotrifluoride, timtec-bb sbb006655, 4-chloro-2-trifluoromethyl benzenisocyanate, pubchem4450, acmc-1c7bp, 4-chloro-2-trifluoromethylphenyl isocyanate, 4-chloro-2-trifluoromethyl-isocyanatobenzene
PubChem CID 2733264
IUPAC Name 4-chloro-1-isocyanato-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=C(C=CC(Cl)=C1)N=C=O

Specifications

Density 1.467
Boiling Point 114°C to 115°C (10 mmHg)
Flash Point 94°C (201°F)
Odor Pungent
Refractive Index 1.501
Quantity 1 g
UN Number UN2206
Beilstein 2214336
Sensitivity Moisture sensitive
Formula Weight 221.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.