missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dimethylphenol, 99%

Product Code. 10530794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10530794 2.5 kg Plastic bottle 2.5kg
10122520 100 g Plastic bottle 100g
10060620 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10530794 Supplier Thermo Scientific Acros Supplier No. 109060025
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88-04-0 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-04-0
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00002324
InChI Key OSDLLIBGSJNGJE-UHFFFAOYSA-N
Synonym chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson
PubChem CID 2723
ChEBI CHEBI:34393
IUPAC Name 4-chloro-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Cl)C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 113.0°C to 116.0°C
Color White to Beige
Boiling Point 246.0°C
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula ClC6H2(CH3)2OH
Quantity 2.5 kg
Beilstein 06, II, 463
Merck Index 15, 2182
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alkali hydroxide solutions,soluble in ethanol,ether and benzene
Formula Weight 156.61
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Chloro-3, 5-dimethylphenol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-3,5-dimethylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.