missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dimethylphenol, 99%

Product Code. p-86815 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10122520

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-04-0 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-04-0
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00002324
InChI Key OSDLLIBGSJNGJE-UHFFFAOYSA-N
Synonym chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson
PubChem CID 2723
ChEBI CHEBI:34393
IUPAC Name 4-chloro-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Cl)C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 113.0°C to 116.0°C
Color White to Beige
Boiling Point 246.0°C
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula ClC6H2(CH3)2OH
Quantity 100 g
Beilstein 06, II, 463
Merck Index 15, 2182
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alkali hydroxide solutions,soluble in ethanol,ether and benzene
Formula Weight 156.61
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Chloro-3, 5-dimethylphenol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-3,5-dimethylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.