missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(chlorosulfonyl)benzoic Acid, TRC

Product Code. 30012900
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012900 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012900 Supplier Toronto Research Chemicals Supplier No. TRCC364950250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2494-79-3
Molecular Formula C7H4Cl2O4S
Molecular Weight (g/mol) 255.08
Synonym 3-(Chlorosulfonyl)-4-chlorobenzoic Acid, 3-Carboxy-6-chlorobenzenesulfonyl Chloride;
IUPAC Name 4-chloro-3-chlorosulfonylbenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)S(=O)(=O)Cl)Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H4Cl2O4S/c8-5-2-1-4(7(10)11)3-6(5)14(9,12)13/h1-3H,(H,10,11)
Chemical Name or Material 4-Chloro-3-(chlorosulfonyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.