missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methoxyaniline, 98%, Thermo Scientific™

Product Code. 13458628 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13458628 25 g 25g
13438628 1 g 1g
13448628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13458628 Supplier Thermo Scientific Acros Supplier No. 448800250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13726-14-2
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.60
MDL Number MFCD00672967
InChI Key LNKBDFVSILQKSI-UHFFFAOYSA-N
Synonym 3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
PubChem CID 13103692
IUPAC Name 4-chloro-3-methoxyaniline
SMILES COC1=C(Cl)C=CC(N)=C1

Specifications

Melting Point 78°C to 81°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(OCH3)NH2
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methoxyaniline
compliance-icons
Product Identifier
  • 4-Chloro-3-methoxyaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.