missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzophenone, 99%

Product Code. 11425437
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425437 25 g 25g
11435437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425437 Supplier Thermo Scientific Alfa Aesar Supplier No. B23451.14
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 56107-02-9 | C13H8ClNO3 | 261.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56107-02-9
Molecular Formula C13H8ClNO3
Molecular Weight (g/mol) 261.66
MDL Number MFCD00007081
InChI Key YBDBYPQFIMSFJW-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
PubChem CID 41743
IUPAC Name (4-chloro-3-nitrophenyl)-phenylmethanone
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1

Specifications

Melting Point 104°C to 106°C
Boiling Point 233°C to 235°C (12 mmHg)
Quantity 25 g
Formula Weight 261.66
Percent Purity 99%
Chemical Name or Material 4-Chloro-3-nitrobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.