missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 10590361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10590361 100 g Glass bottle 100g
10448060 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10590361 Supplier Thermo Scientific Acros Supplier No. 109701000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 121-17-5
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.55
MDL Number MFCD00007084
InChI Key TZGFQIXRVUHDLE-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride
PubChem CID 8462
IUPAC Name 1-chloro-2-nitro-4-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2.0°C
Color Yellow
Density 1.5110g/mL
Boiling Point 222.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(NO2)CF3
Refractive Index 1.4883 to 1.4903
Quantity 100 g
Specific Gravity 1.511
Solubility Information Solubility in water: 660mg/l water (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 225.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-nitrobenzotrifluoride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-3-nitrobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.