missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrocinnamic acid, 98%

Product Code. 11467147
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467147 5 g 5g
11477147 25 g 25g
11487147 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467147 Supplier Thermo Scientific Alfa Aesar Supplier No. A14954.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 20797-48-2 | C9H6ClNO4 | 227.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20797-48-2
Molecular Formula C9H6ClNO4
Molecular Weight (g/mol) 227.6
MDL Number MFCD00063311
InChI Key QBDALTIMHOITIU-DUXPYHPUSA-N
Synonym 4-chloro-3-nitrocinnamic acid, trans-4-chloro-3-nitrocinnamic acid, 3-4-chloro-3-nitrophenyl acrylic acid, e-3-4-chloro-3-nitrophenyl acrylic acid, 2e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, 4-chloro-3-nitro-cinnamic acid, 3-nitro-4-chlorocinnamic acid, qbdaltimhoitiu-duxpyhpusa, 3-4-chloro-3-nitrophenyl propenoic acid
PubChem CID 688108
SMILES C1=CC(=C(C=C1C=CC(=O)O)[N+](=O)[O-])Cl

Specifications

Melting Point 186°C to 188°C
Quantity 5 g
Formula Weight 227.61
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrocinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.